2-Amino-2-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)propanamide hydrochloride
Supplier: Matrix Scientific
053552-2.5G
053552-500MG
101904-096EA
838.46
USD
101904-096
101904-094
2-Amino-2-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)propanamide hydrochloride
2-Amino-2-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₁ClN₂O₂ |
MDL Number:
MFCD13562869 CAS Number: 1220021-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...