5-Bromo-3-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyridinamine

Supplier: Matrix Scientific

053544-500MG 053544-2.5G
101904-062EA 364.62 USD
101904-062 101904-064
5-Bromo-3-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyridinamine
5-Bromo-3-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₇BrN₂O
MDL Number: MFCD13562861
CAS Number: 1220029-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR