3-Amino-N-(4-methoxyphenyl)propanamide hydrochloride

Supplier: Matrix Scientific

053524-500MG 053524-2.5G
101903-982EA 364.62 USD
101903-982 101903-984
3-Amino-N-(4-methoxyphenyl)propanamide hydrochloride
3-Amino-N-(4-methoxyphenyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O₂
MDL Number: MFCD13562843
CAS Number: 57500-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR