5-Bromo-N-(4-methoxyphenyl)-2-pyridinamine
Supplier: Matrix Scientific
053519-2.5G
053519-500MG
101903-964EA
838.46
USD
101903-964
101903-962
5-Bromo-N-(4-methoxyphenyl)-2-pyridinamine
5-Bromo-N-(4-methoxyphenyl)-2-pyridinamine
Hazard: Irritant
Formula:
C₁₂H₁₁BrN₂O |
MDL Number:
MFCD13562839 CAS Number: 1220020-51-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...