3-Amino-N-(4-fluorophenyl)propanamide hydrochloride
Supplier: Matrix Scientific
053499-2.5G
053499-500MG
101903-882EA
364.62
USD
101903-882
101903-884
3-Amino-N-(4-fluorophenyl)propanamide hydrochloride
3-Amino-N-(4-fluorophenyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₂ClFN₂O |
MDL Number:
MFCD13193985 CAS Number: 938515-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...