6-Chloro-N-(4-fluorophenyl)-4-pyrimidinamine

Supplier: Matrix Scientific

053486-500MG 053486-2.5G
101903-832EA 838.46 USD
101903-832 101903-830
6-Chloro-N-(4-fluorophenyl)-4-pyrimidinamine
6-Chloro-N-(4-fluorophenyl)-4-pyrimidinamine

Hazard: Irritant

Formula: C₁₀H₇ClFN₃
MDL Number: MFCD09756978
CAS Number: 945896-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR