3-Amino-4-chloro-N-(4-pyridinylmethyl)benzenesulfonamide
Supplier: Matrix Scientific
053482-2.5G
053482-500MG
101903-814EA
364.62
USD
101903-814
101903-816
3-Amino-4-chloro-N-(4-pyridinylmethyl)benzenesulfonamide
3-Amino-4-chloro-N-(4-pyridinylmethyl)benzenesulfonamide
Hazard: Irritant
Formula:
C₁₂H₁₂ClN₃O₂S |
MDL Number:
MFCD12618500 CAS Number: 1040334-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...