N-(4-Pyridinylmethyl)-4-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053480-2.5G
053480-500MG
101903-808EA
838.46
USD
101903-808
101903-806
N-(4-Pyridinylmethyl)-4-piperidinecarboxamide hydrochloride
N-(4-Pyridinylmethyl)-4-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₈ClN₃O |
MDL Number:
MFCD13562814 CAS Number: 1219976-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...