2-Amino-2-methyl-N-(4-pyridinylmethyl)propanamide hydrochloride
Supplier: Matrix Scientific
053475-2.5G
053475-500MG
101903-788EA
838.46
USD
101903-788
101903-786
2-Amino-2-methyl-N-(4-pyridinylmethyl)propanamide hydrochloride
2-Amino-2-methyl-N-(4-pyridinylmethyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₆ClN₃O |
MDL Number:
MFCD13562809 CAS Number: 1220035-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...