2-[(4-Pyridinylmethyl)amino]nicotinic acid
Supplier: Matrix Scientific
053464-2.5G
053464-500MG
101903-744EA
838.46
USD
101903-744
101903-742
2-[(4-Pyridinylmethyl)amino]nicotinic acid
2-[(4-Pyridinylmethyl)amino]nicotinic acid
Hazard: Irritant
Formula:
C₁₂H₁₁N₃O₂ |
MDL Number:
MFCD11118784 CAS Number: 854382-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...