2-Chloro-N-(2-pyridinylmethyl)nicotinamide
Supplier: Matrix Scientific
053440-2.5G
053440-500MG
101903-646EA
364.62
USD
101903-646
101903-648
2-Chloro-N-(2-pyridinylmethyl)nicotinamide
2-Chloro-N-(2-pyridinylmethyl)nicotinamide
Hazard: Irritant
Formula:
C₁₂H₁₀ClN₃O |
MDL Number:
MFCD01351665 CAS Number: 550314-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...