6-Chloro-N-(2-pyridinylmethyl)-4-pyrimidinamine
Supplier: Matrix Scientific
053434-500MG
053434-2.5G
101903-624EA
340.05
USD
101903-624
101903-622
6-Chloro-N-(2-pyridinylmethyl)-4-pyrimidinamine
6-Chloro-N-(2-pyridinylmethyl)-4-pyrimidinamine
Hazard: Irritant
Formula:
C₁₀H₉ClN₄ |
MDL Number:
MFCD13562783 CAS Number: 1125865-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...