3-Amino-4-chloro-N-(3-pyridinylmethyl)benzenesulfonamide
Supplier: Matrix Scientific
053430-2.5G
053430-500MG
101903-606EA
364.62
USD
101903-606
101903-608
3-Amino-4-chloro-N-(3-pyridinylmethyl)benzenesulfonamide
3-Amino-4-chloro-N-(3-pyridinylmethyl)benzenesulfonamide
Hazard: Irritant
Formula:
C₁₂H₁₂ClN₃O₂S |
MDL Number:
MFCD12615241 CAS Number: 1036458-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...