N-(3-Pyridinylmethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride

Supplier: Matrix Scientific

053429-2.5G 053429-500MG
101903-602EA 364.62 USD
101903-602 101903-604
N-(3-Pyridinylmethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
N-(3-Pyridinylmethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₆ClN₅O
MDL Number: MFCD13562780
CAS Number: 1220038-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR