N-(3-Pyridinylmethyl)-4-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053428-2.5G
053428-500MG
101903-598EA
364.62
USD
101903-598
101903-600
N-(3-Pyridinylmethyl)-4-piperidinecarboxamide hydrochloride
N-(3-Pyridinylmethyl)-4-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₈ClN₃O |
MDL Number:
MFCD13562779 CAS Number: 1220035-36-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...