6-Chloro-N-(3-pyridinylmethyl)-2-pyrazinamine
Supplier: Matrix Scientific
053410-500MG
053410-2.5G
101903-528EA
838.46
USD
101903-528
101903-526
6-Chloro-N-(3-pyridinylmethyl)-2-pyrazinamine
6-Chloro-N-(3-pyridinylmethyl)-2-pyrazinamine
Hazard: Irritant
Formula:
C₁₀H₉ClN₄ |
MDL Number:
MFCD13562767 CAS Number: 629657-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...