6-Chloro-N-(3-pyridinylmethyl)-4-pyrimidinamine

Supplier: Matrix Scientific

053408-500MG 053408-2.5G
101903-518EA 149.23 USD
101903-518 101903-520
6-Chloro-N-(3-pyridinylmethyl)-4-pyrimidinamine
6-Chloro-N-(3-pyridinylmethyl)-4-pyrimidinamine

Hazard: Irritant

Formula: C₁₀H₉ClN₄
MDL Number: MFCD13562766
CAS Number: 1219957-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR