5-Bromo-4-methyl-N-(3-pyridinylmethyl)-2-pyridinamine
Supplier: Matrix Scientific
053407-2.5G
053407-500MG
101903-514EA
364.62
USD
101903-514
101903-516
5-Bromo-4-methyl-N-(3-pyridinylmethyl)-2-pyridinamine
5-Bromo-4-methyl-N-(3-pyridinylmethyl)-2-pyridinamine
Hazard: Irritant
Formula:
C₁₂H₁₂BrN₃ |
MDL Number:
MFCD13562765 CAS Number: 1220038-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...