N-Benzyl-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053401-500MG 053401-2.5G
101903-490EA 461.54 USD
101903-490 101903-492
N-Benzyl-3-piperidinecarboxamide hydrochloride
N-Benzyl-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O
MDL Number: MFCD13562762
CAS Number: 668461-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR