N-Benzyl-6-chloro-2-pyridinamine

Supplier: Matrix Scientific

053391-500MG 053391-2.5G
101903-450EA 364.62 USD
101903-450 101903-452
N-Benzyl-6-chloro-2-pyridinamine
N-Benzyl-6-chloro-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₁ClN₂
MDL Number: MFCD04625566
CAS Number: 29449-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR