2-(Benzylamino)-5-bromopyridine
053390-2.5G
101903-448EA
838.46
USD
101903-448
2-(Benzylamino)-5-bromopyridine
2-(Benzylamino)-5-bromopyridine
Hazard: Irritant
Formula:
C₁₂H₁₁BrN₂ |
MDL Number:
MFCD04215647 CAS Number: 280116-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...