N-Benzyl-5-bromo-4-methyl-2-pyridinamine

Supplier: Matrix Scientific

053382-2.5G 053382-500MG
101903-414EA 364.62 USD
101903-414 101903-416
N-Benzyl-5-bromo-4-methyl-2-pyridinamine
N-Benzyl-5-bromo-4-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₃H₁₃BrN₂
MDL Number: MFCD13562753
CAS Number: 1219964-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR