N-(4-Methylphenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
Supplier: Matrix Scientific
053378-500MG
053378-2.5G
101903-398EA
364.62
USD
101903-398
101903-400
N-(4-Methylphenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
N-(4-Methylphenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₄H₁₇ClN₄O MW: 292.77 g/mol |
MDL Number:
MFCD13562751 CAS Number: 1219976-41-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...