N-(4-Methylphenyl)-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053375-500MG 053375-2.5G
101903-386EA 364.62 USD
101903-386 101903-388
N-(4-Methylphenyl)-2-piperidinecarboxamide hydrochloride
N-(4-Methylphenyl)-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O
MDL Number: MFCD13562748
CAS Number: 1236263-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR