2-Amino-N-(4-methylphenyl)acetamide hydrochloride
Supplier: Matrix Scientific
053367-2.5G
053367-500MG
101903-354EA
364.62
USD
101903-354
101903-356
2-Amino-N-(4-methylphenyl)acetamide hydrochloride
2-Amino-N-(4-methylphenyl)acetamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₃ClN₂O |
MDL Number:
MFCD13562742 CAS Number: 54643-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...