N2-(4-Methylphenyl)-2,3-pyridinediamine

Supplier: Matrix Scientific

053362-500MG 053362-2.5G
101903-334EA 364.62 USD
101903-334 101903-336
N2-(4-Methylphenyl)-2,3-pyridinediamine
N2-(4-Methylphenyl)-2,3-pyridinediamine

Hazard: Irritant

Formula: C₁₂H₁₃N₃
MDL Number: MFCD11101070
CAS Number: 70358-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR