3-Amino-4-chloro-N-(3-methylphenyl)benzenesulfonamide

053353-2.5G
101903-300EA 838.46 USD
101903-300
3-Amino-4-chloro-N-(3-methylphenyl)benzenesulfonamide
3-Amino-4-chloro-N-(3-methylphenyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₃H₁₃ClN₂O₂S
MW: 296.78 g/mol
MDL Number: MFCD12440297
CAS Number: 1036624-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR