6-Chloro-N-(3-methylphenyl)-2-pyridinamine

Supplier: Matrix Scientific

053340-500MG 053340-2.5G
101903-246EA 364.62 USD
101903-246 101903-248
6-Chloro-N-(3-methylphenyl)-2-pyridinamine
6-Chloro-N-(3-methylphenyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₁ClN₂
MDL Number: MFCD13562723
CAS Number: 1220019-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR