3-Chloro-N-(3-methylphenyl)-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

053332-500MG 053332-2.5G
101903-216EA 838.46 USD
101903-216 101903-214
3-Chloro-N-(3-methylphenyl)-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-(3-methylphenyl)-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₃H₁₀ClF₃N₂
MDL Number: MFCD13562719
CAS Number: 1219957-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR