N-(2-Methylphenyl)-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053325-500MG 053325-2.5G
101903-186EA 364.62 USD
101903-186 101903-188
N-(2-Methylphenyl)-4-piperidinecarboxamide hydrochloride
N-(2-Methylphenyl)-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O
MDL Number: MFCD13562714
CAS Number: 1219976-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR