Ethyl-3-amino-4-(2-toluidino)benzoate

Supplier: Matrix Scientific

053302-500MG
101903-094EA 364.62 USD
101903-094
Ethyl-3-amino-4-(2-toluidino)benzoate
Ethyl-3-amino-4-(2-toluidino)benzoate

Hazard: Irritant

Formula: C₁₆H₁₈N₂O₂
MDL Number: MFCD13562701
CAS Number: 1220035-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR