N-(3-Ethoxypropyl)-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053297-500MG 053297-2.5G
101903-076EA 838.46 USD
101903-076 101903-074
N-(3-Ethoxypropyl)-2-piperidinecarboxamide hydrochloride
N-(3-Ethoxypropyl)-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃ClN₂O₂
MDL Number: MFCD13562697
CAS Number: 1236261-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR