2-Amino-N-(3-ethoxypropyl)acetamide hydrochloride
Supplier: Matrix Scientific
053289-2.5G
053289-500MG
101903-044EA
838.46
USD
101903-044
101903-042
2-Amino-N-(3-ethoxypropyl)acetamide hydrochloride
2-Amino-N-(3-ethoxypropyl)acetamide hydrochloride
Hazard: Irritant
Formula:
C₇H₁₇ClN₂O₂ |
MDL Number:
MFCD13562689 CAS Number: 1219964-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...