N-[3-(Dimethylamino)propyl]-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide dihydrochloride

053274-2.5G
101902-984EA 838.46 USD
101902-984
N-[3-(Dimethylamino)propyl]-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide dihydrochloride
N-[3-(Dimethylamino)propyl]-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₃Cl₂N₅O
MW: 324.26 g/mol
MDL Number: MFCD13562682
CAS Number: 1220038-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR