2-Amino-N-cyclohexylpropanamide hydrochloride

053238-2.5G
101902-840EA 838.46 USD
101902-840
2-Amino-N-cyclohexylpropanamide hydrochloride
2-Amino-N-cyclohexylpropanamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O
MDL Number: MFCD13562648
CAS Number: 34582-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR