2-(Cyclohexylamino)isonicotinic acid
Supplier: Matrix Scientific
053230-2.5G
053230-500MG
101902-808EA
364.62
USD
101902-808
101902-810
2-(Cyclohexylamino)isonicotinic acid
2-(Cyclohexylamino)isonicotinic acid
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₂ |
MDL Number:
MFCD11131952 CAS Number: 1019461-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...