5-Bromo-3-methyl-N-(tetrahydro-2-furanylmethyl)-2-pyridinamine
Supplier: Matrix Scientific
053208-500MG
053208-2.5G
101902-722EA
838.46
USD
101902-722
101902-720
5-Bromo-3-methyl-N-(tetrahydro-2-furanylmethyl)-2-pyridinamine
5-Bromo-3-methyl-N-(tetrahydro-2-furanylmethyl)-2-pyridinamine
Hazard: Irritant
Formula:
C₁₁H₁₅BrN₂O |
MDL Number:
MFCD13562631 CAS Number: 1220017-11-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...