6-Chloro-N-(tetrahydro-2-furanylmethyl)-2-pyrazinamine
Supplier: Matrix Scientific
053203-2.5G
053203-500MG
101902-702EA
838.46
USD
101902-702
101902-700
6-Chloro-N-(tetrahydro-2-furanylmethyl)-2-pyrazinamine
6-Chloro-N-(tetrahydro-2-furanylmethyl)-2-pyrazinamine
Hazard: Irritant
Formula:
C₉H₁₂ClN₃O |
MDL Number:
MFCD13562630 CAS Number: 1220037-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...