3-Nitro-4-[(tetrahydro-2-furanylmethyl)amino]benzoic acid

Supplier: Matrix Scientific

053199-500MG 053199-2.5G
101902-684EA 364.62 USD
101902-684 101902-686
3-Nitro-4-[(tetrahydro-2-furanylmethyl)amino]benzoic acid
3-Nitro-4-[(tetrahydro-2-furanylmethyl)amino]benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₅
MDL Number: MFCD02111672
CAS Number: 309942-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR