N-(2-Furylmethyl)-2-pyrrolidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053192-2.5G
053192-500MG
101902-658EA
838.46
USD
101902-658
101902-656
N-(2-Furylmethyl)-2-pyrrolidinecarboxamide hydrochloride
N-(2-Furylmethyl)-2-pyrrolidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₅ClN₂O₂ |
MDL Number:
MFCD09971403 CAS Number: 1078162-92-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...