3-Amino-N-(2-furylmethyl)propanamide hydrochloride
Supplier: Matrix Scientific
053188-2.5G
053188-500MG
101902-640EA
364.62
USD
101902-640
101902-642
3-Amino-N-(2-furylmethyl)propanamide hydrochloride
3-Amino-N-(2-furylmethyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₃ClN₂O₂ |
MDL Number:
MFCD13562620 CAS Number: 1220035-20-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...