N-(5-Bromo-3-methyl-2-pyridinyl)-N-(2-furylmethyl)amine
Supplier: Matrix Scientific
053182-2.5G
053182-500MG
101902-618EA
838.46
USD
101902-618
101902-616
N-(5-Bromo-3-methyl-2-pyridinyl)-N-(2-furylmethyl)amine
N-(5-Bromo-3-methyl-2-pyridinyl)-N-(2-furylmethyl)amine
Hazard: Irritant
Formula:
C₁₁H₁₁BrN₂O |
MDL Number:
MFCD13562617 CAS Number: 1220028-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...