2-[(2-Furylmethyl)amino]isonicotinic acid
Supplier: Matrix Scientific
053178-2.5G
053178-500MG
101902-602EA
838.46
USD
101902-602
101902-600
2-[(2-Furylmethyl)amino]isonicotinic acid
2-[(2-Furylmethyl)amino]isonicotinic acid
Hazard: Irritant
Formula:
C₁₁H₁₀N₂O₃ |
MDL Number:
MFCD11131988 CAS Number: 1019387-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...