2-Amino-N-(3-methoxypropyl)-2-methylpropanamide hydrochloride
Supplier: Matrix Scientific
053138-2.5G
053138-500MG
101902-442EA
838.46
USD
101902-442
101902-440
2-Amino-N-(3-methoxypropyl)-2-methylpropanamide hydrochloride
2-Amino-N-(3-methoxypropyl)-2-methylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₉ClN₂O₂ |
MDL Number:
MFCD13562598 CAS Number: 1219963-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...