2-Amino-N-(3-methoxypropyl)-3-methylbutanamide hydrochloride
Supplier: Matrix Scientific
053137-2.5G
053137-500MG
101902-438EA
838.46
USD
101902-438
101902-436
2-Amino-N-(3-methoxypropyl)-3-methylbutanamide hydrochloride
2-Amino-N-(3-methoxypropyl)-3-methylbutanamide hydrochloride
Hazard: Irritant
Formula:
C₉H₂₁ClN₂O₂ |
MDL Number:
MFCD13562597 CAS Number: 1236256-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...