3-Amino-N-(3-methoxypropyl)propanamide hydrochloride
Supplier: Matrix Scientific
053136-2.5G
053136-500MG
101902-432EA
364.62
USD
101902-432
101902-434
3-Amino-N-(3-methoxypropyl)propanamide hydrochloride
3-Amino-N-(3-methoxypropyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₇H₁₇ClN₂O₂ |
MDL Number:
MFCD13562596 CAS Number: 1220037-58-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...