2-Amino-N-(3-methoxypropyl)acetamide hydrochloride

Supplier: Matrix Scientific

053133-500MG 053133-2.5G
101902-420EA 364.62 USD
101902-420 101902-422
2-Amino-N-(3-methoxypropyl)acetamide hydrochloride
2-Amino-N-(3-methoxypropyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₅ClN₂O₂
MDL Number: MFCD13562593
CAS Number: 1219964-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR