5-Bromo-N-(3-methoxypropyl)-3-methyl-2-pyridinamine
Supplier: Matrix Scientific
053130-500MG
053130-2.5G
101902-410EA
838.46
USD
101902-410
101902-408
5-Bromo-N-(3-methoxypropyl)-3-methyl-2-pyridinamine
5-Bromo-N-(3-methoxypropyl)-3-methyl-2-pyridinamine
Hazard: Irritant
Formula:
C₁₀H₁₅BrN₂O |
MDL Number:
MFCD13562591 CAS Number: 1220035-09-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...