2-[(3-Methoxypropyl)amino]isonicotinic acid
Supplier: Matrix Scientific
053126-500MG
053126-2.5G
101902-394EA
838.46
USD
101902-394
101902-392
2-[(3-Methoxypropyl)amino]isonicotinic acid
2-[(3-Methoxypropyl)amino]isonicotinic acid
Hazard: Irritant
Formula:
C₁₀H₁₄N₂O₃ |
MDL Number:
MFCD11204937 CAS Number: 1094292-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...