2-Amino-N-(2-hydroxy-1,1-dimethylethyl)-3-phenylpropanamide hydrochloride
Supplier: Matrix Scientific
053113-500MG
053113-2.5G
101902-342EA
838.46
USD
101902-342
101902-340
2-Amino-N-(2-hydroxy-1,1-dimethylethyl)-3-phenylpropanamide hydrochloride
2-Amino-N-(2-hydroxy-1,1-dimethylethyl)-3-phenylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₁ClN₂O₂ |
MDL Number:
MFCD13562585 CAS Number: 1236261-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...